Ameisensäure ≥88,0% (durch titrimetrische Analyse), TraceSELECT™ für die Spurenanalyse, Fluka™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
06454-250ML
06454-250MLEA 315.49 EUR InStock
06454-250ML
Ameisensäure ≥88,0% (durch titrimetrische Analyse), TraceSELECT™ für die Spurenanalyse, Fluka™
Ameisensäure
TraceSELECT™ Ultra and TraceSELECT
  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
Formel: HCO₂H
Siedepunkt: 100,4 °C (1.013 hPa)
Melting Pt: 4 °C
Dichte: 1,22 g/cm³ (20 °C)
Flash-Pt: 49,5 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003297
CAS-Nummer: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
REACH: 01-2119491174-37
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=e79a8c7fb6c358f57ed533653859d68b28119045&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Assay Min. 88.0 %
Density (D 20/4) 1.190 - 1.220
Non-volatile matter Max. 0.0001 %
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 1 ppb
Barium (Ba) Max. 1 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 20 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.5 ppb
Cobalt (Co) Max. 1 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 10 ppb
Gallium (Ga) Max. 0.5 ppb
Mercury (Hg) Max. 1 ppb
Indium (In) Max. 0.5 ppb
Iridium (Ir) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 5 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 10 ppb
Nickel (Ni) Max. 2 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 0.5 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 1 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 1 ppb
Zinc (Zn) Max. 5 ppb
Zirconium (Zr) Max. 0.5 ppb
Chloride (Cl) Max. 10 ppm
Sulfate (SO₄) Max. 0.5 ppm
Ammonia (NH₃) Max. 10 ppm
Acetic acid (CH₃COOH) Max. 0.05 %
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge