Organic reference standard, Octyl gallate, analytical standard, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
06717-500MG
06717-500MGEA 76.2 EUR InStock
06717-500MG
Organic reference standard, Octyl gallate, analytical standard, Supelco®
Standards Organische Standards
Octyl gallate is an ester of gallic acid which is generally used as an antioxidant in food and pharmaceutical industries. It can act against the oxidative damage caused by reactive oxygen species (ROS) in the form of hydroxyl radicals or hydrogen peroxides, and reactive sulphur species (RSS).
Octyl gallate can be used as a reference standard for the determination of octyl gallate in edible vegetable oil by high performance liquid chromatography/time-of-flight mass spectrometry (HPLC/TOF-MS) as well as alternating penalty trilinear decomposition (APTLD) combined with HPLC-diode array detection (DAD) dry foods by reversed-phase HPLC with photodiode array (PDA) detection.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88761bf7be621564d4b69abff5cb06dadd48cae1&vid=88761bf7be621564d4b69abff5cb06dadd48cae1&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge