Butyrate standard for IC, TraceCERT®, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
08089-100ML
08089-100MLEA 65.4 EUR InStock
08089-100ML
Butyrate standard for IC, TraceCERT®, Supelco®
Standards Anionische Standards Butyrate Standards
Prepared with sodium butyrate and water; filtered through a 0,2 μm membrane filter and stabilised with sodium azide.
Zertifizierungen: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge