VWR®, Hybridisation buffers with 50% formamide
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0973-50MLEA 254 EUR InStock
0973-50ML
VWR®, Hybridisation buffers with 50% formamide
Puffer Elektrophorese-Puffer Hybridisierungspuffer
Hybridisation cocktail offering blocking and hybridisation for optimised complimentary base pairing of DNA and RNA in Southern blot, Northern blot, dot blot, colony and in situ hybridisation.
Achtung! Bitte beachten: Store frozen.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge