Granular dessicant, Sicapent® with indicator, Supelco®
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.00543.0500 1.00543.2800
1.00543.0500EA 92.2 EUR InStock
1.00543.0500 1.00543.2800
Granular dessicant, Sicapent® with indicator, Supelco®
Exsikkatoren Trockenmittel
Sicapent is a granular dessicant comprising ~75% phoshorus pentoxide on an inert inorganic carrier.
  • Ease of use
  • Fast water adsorption due to large surface area
  • Colour indicator - colourless to green to blue
  • Remains in particle form after 100% water uptake
Appliations include drying liquids, filling drying tubes, particularly for fast flowing gases in drying tubes.
Bulk density: ~300 g/L
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge