Media fill-sterility test for pharmaceutical industries
Lieferant: MERCK MILLIPORE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.00800.5000 1.00800.0500 1.00550.5000 1.00550.0500
1.00800.5000EA 1540 EUR InStock
1.00800.5000 1.00800.0500 1.00550.5000 1.00550.0500
Media fill-sterility test for pharmaceutical industries
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Be assured that Merck offers the quality TSB that you are looking for in your aseptic process simulation.
  • Triple bagged and ready for cleanroom use: Three layers of protection allow you to add directly to production vessels and delivery systems.
  • Irradiation process using 48-68 kGy gamma: Robust validated process ensures that media related positives are eliminated from your aseptic process simulation. The irradiation process inactivates any viable bacteria, yeast, mold, spores and Mycoplasma.
  • Every lot is Mycoplasma tested: Validated PCR technology QC testing process for Mycoplasma detection to ensure absence of this difficult contaminant.
  • Growth promotion and sterility tested exceeding expectations of USP/EP standards
  • Produced and tested according to ISO standard 11133.
  • Complete traceability documentation for compliance with BSE/TSE requirements.
  • Excellent filterability and solubility reduces preparation time and prolongs filter life during media fills when compared to traditional powdered media, thus saving time and money by reducing filter changes.
  • Exclusive granulated format
  • Reduction of dust during preparation, lower risk to your operators. Easier handling and less dust to adhere to equipment.
When you are performing a media fill test, the last thing that should be on your mind is if your TSB media is going to affect your validated process.
Every lot Merck Millipore’s irradiated media is tested for Sterility, absence of Mycoplasma, and comes in the high-performing granulated format. Along with the cleanroom ready triple-wrapped packaging, the advantages are clear.
Zertifizierungen: In compliance with DIN ISO 4796-1 standards.
Lieferumfang: Supplied with screw cap and pouring ring.
Verpackung: The bottles are packaged in shrinked-wrapped modules.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..