Brillantkresylblau -Lösung biologische Färbemittel, für die Mikroskopie, Sigma-Aldrich®
Lieferant: MERCK MILLIPORE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.01384.0100 1.01384.9025
1.01384.0100EA 51.1 EUR InStock
1.01384.0100 MERC1.01384.9025
Brillantkresylblau -Lösung biologische Färbemittel, für die Mikroskopie, Sigma-Aldrich®
Brillantkresylblau
The Brilliant cresyl blue solution is used in hematology as reagent for the staining of blood for the subsequent manual counting of blood cells. The regenerative capacity of erythrocytes can be monitored with the ratio of reticulocytes. In addition, the determination of the Substantia granulo-filamentosa (ribonucleoproteins) is possible with fresh, unfixed young erythrocytes (in a supra-vitral staining).
The Brilliant cresyl blue solution is an IVD & CE product and 100 ml of the solution are capable of staining up to 5000 slides.
Formel: C₃₄H₄₂Cl₄N₈O₂Zn
Dichte: 1,01 g/cm3 (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00148901
CAS-Nummer: 81029-05-2
IVDLogo
Order Now
Spezifikation Testergebnisse
Application test (Reticulocyte staining) passes test
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge