Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bariumfluorid, wasserfrei ≥99,995%, Pulver, Optipur®

Lieferant: MERCK MILLIPORE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

1.01705.9050
1.01705.9050EA 0 EUR InStock
1.01705.9050
Bariumfluorid, wasserfrei ≥99,995%, Pulver, Optipur®
Barium fluoride
Optipur® materials are inorganic compounds, primarily metal halides, and available as anhydrous powders, beads or pieces for immediate usability and easy handling. They are of the highest purity, and specifically suited for the production of single crystals, used in demanding optical applications or scintillators, for example medical imaging and radiation detection.

  • High purity
  • Anhydrous powders or beads
  • Mainly metal halides
  • Immediate usability and easy handling
  • Special packaging for hygroscopic materials

The purity of each production batch is strictly controlled with careful attention paid to impurities that have an influence on transmission, light scattering, or scintillation efficiency. Hygroscopic materials are typically vacuum-dried at high temperature and are packaged for dry storage.


Formel: BaF₂
Siedepunkt: 2260 °C (1013 hPa)
Melting Pt: 1353 °C
Dichte: 4,89 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003450
CAS-Nummer: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge