Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Neo-Cytocolor® Quick Staining Kit for cytological samples, Sigma-Aldrich®

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

1.01971.0001
1.01971.0001EA 0 EUR InStock
1.01971.0001
Neo-Cytocolor® Quick Staining Kit for cytological samples, Sigma-Aldrich®
Neo-Cytocolor® Quick Staining Kit for cytological samples
For gynecological and clinical specimens. Contains Neo-Clear®, xylene substitute

Neo-Cytocolor® is a quick - 4 minute - staining kit based on the standard staining method acc. to Szczepanik for use in the analysis of cytological specimens. This kit dispenses with the use of xylene, containing the xylene substitute NEO-CLEAR® instead. Standard fixatives can be used. Neo-Cytocolor® is an IVD product. As it also is CE-certified, it can be applied in diagnostics in an accredited laboratory.The package is sufficient for approx. 1000 applications.

Specifications
Suitability for microscopy (Vaginal smear): passes test
Nuclei: blue to dark violet
Cyanophilic cytoplasm: blue-green
Eosinophilic cytoplasm: pink
keratinized cytoplasm: pink-orange
Microorganisms: blue-violet
Erythrocytes: red

The package (6x 500 ml) is sufficient for approx. 1000 applications.


UN: 1219
ADR: 3 II
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge