Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

1-Butyl-2-pyrrolidone, EMPLURA®

Lieferant: MERCK MILLIPORE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

1.03818.1000 1.03818.2500 1.03818.6025
1.03818.1000EA 65.7 EUR InStock
1.03818.1000 1.03818.2500 1.03818.6025
1-Butyl-2-pyrrolidone, EMPLURA®
1-Butyl-2-pyrrolidone
1-Butylpyrrolidin-2-one is a safer alternative to N-Methyl-2-pyrrolidone (NMP), N,N-Dimethylacetamide (DMA), Dimethyl sulfoxide (DMSO) and N,N-Dimethylformamide (DMF), which face increasing regulatory pressure. As opposed to NMP, DMF and DMA, 1-Butylpyrrolidin-2-one is not classified as developmentally reprotoxic.

  • Excellent solvency power and water miscibility
  • High boiling point
  • High chemical and thermal stability
  • Not classified as a developmentalor geno-toxin
  • Inherently bio-degradable
  • Lower volatility compared to NMP
  • Reliable alternative for REACH-restricted DMA, DMF, NMP


Formel: C₈H₁₅NO
MW: 141.21 g/mol
MDL Number: MFCD00020876
CAS-Nummer: 3470-98-2
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge