Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Leishman's Eosin-Methylenblau modifizierte Lösung Färbemittel für Histologie, Sigma-Aldrich®

Lieferant: MERCK MILLIPORE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

1.05387.0500 1.05387.0000 1.05387.1022
1.05387.0500EA 103 EUR InStock
1.05387.0500 1.05387.0000 1.05387.1022
Leishman's Eosin-Methylenblau modifizierte Lösung Färbemittel für Histologie, Sigma-Aldrich®
Leishman's Eosin-Methylenblau
For staining of blood and bone marrow smears and clinical-cytological specimens

The staining method according to Leishman is one of the standard techniques in hematological diagnostic procedures. Like Wright's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The typical resulting color of cell nuclei is red to purple, which depends on the buffer tablets (Buffer tablets pH 7.2 or Buffer tablets pH 6.8). 500 mL of the IVD & CE-registered Leishman's eosin methylene blue solution are sufficient for staining up to 1600 slides in Hellendahl cells. It is registered as IVD and CE product.

Preparative capacity: 1000 ml

3200 slides automate/staining cell (diluted)
500 slides staining rack (undiluted)


Siedepunkt: 65 °C (1013 hPa)
Dichte: 0,79 g/cm3 (20 °C)
Flash-Pt: 12 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00131498
CAS-Nummer: 12627-53-1
UN: 1230
ADR: 3 (6.1),II
IVDLogo
Order Now
Spezifikation Testergebnisse
Suitability for microscopy (Blood smear) passes test
Erythrocytes reddish
Nuclei violet
Eosinophilic granules brick-red
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge