Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Natriumbenzoat 99,0-100,5%, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E211, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

1.06290.1000 1.06290.9029 1.06290.9525 1.06290.9999
1.06290.1000EA 225 EUR InStock
1.06290.1000 1.06290.9029 1.06290.9525 1.06290.9999
Natriumbenzoat 99,0-100,5%, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E211, SAFC®
Natriumbenzoat
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*). Corresponds to Ph Eur,BP,NF,FCC,E 211

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur,BP,NF,FCC,E 211


Formel: C₆H₅COONa
Melting Pt: 410…430 °C
Dichte: 1,44 g/cm³ (20 °C)
Flash-Pt: >100 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00012463
CAS-Nummer: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654
Order Now
Spezifikation Testergebnisse
Assay (acidimetric, calc. on anhydrous substance) 99.0 - 100.5 %
Assay (HPLC, calc. on anhydrous substance) 99.0 - 101.0 %
Identity passes test
Appearance of solution (100 g/l, water) passes test
Acidity or alkalinity passes test
Chloride (Cl) ≤ 0.02 %
Sulphate (SO₄) ≤ 0.01 %
Total chlorine ≤ 0.03 %
Heavy metals (as Pb) ≤ 10 ppm
As (Arsenic) ≤ 1 ppm
Cd (Cadmium) ≤ 10 ppm
Cu (Copper) ≤ 10 ppm
Hg (Mercury) ≤ 1 ppm
Pb (Lead) ≤ 2 ppm
Zn (Zinc) ≤ 10 ppm
Benzene ≤ 2 ppm
Toluene ≤ 890 ppm
Other residual solvents (ICH Q3C) excluded by production process
Multinuclear acids passes test
Oxidizable matter passes test
Water (according to Karl Fischer) ≤ 1.5 %
Corresponds to Ph. Eur., BP, NF, FCC, E211

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge