Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Natriumfluorid, EMPROVE® API USP, Ph. Eur., BP suitable for use as an active pharmaceutical ingredient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

1.06441.1000 1.06441.9025
1.06441.1000EA 696 EUR InStock
1.06441.1000 1.06441.9025
Natriumfluorid, EMPROVE® API USP, Ph. Eur., BP suitable for use as an active pharmaceutical ingredient, SAFC®
Sodium fluoride
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Zertifizierungen: Conforms to Ph. Eur., BP, USP


Formel: NaF
MW: 41.99 g/mol
Siedepunkt: 1700
Melting Pt: 996 °C
Dichte: 2.8 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003524
CAS-Nummer: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 13,08691
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=5de299c7c1002dab6bcd571d3b9397558a27a5dc&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Assay (Precipitative Titration, Calculated on Dried Substance) 98.5 - 100.5%
Identity Passes Test
Appearance of solution Clear and Colourless
Alkalinity (as NaOH) ≤0.1%
Acidity (as HF) ≤0.2%
Chloride (Cl) ≤0.01%
Hexafluorosilicate (SiF₆) ≤0.1%
Sulfate (SO₄) ≤0.02%
Heavy metals (as Pb) ≤0.003%
Fe (Iron) ≤0.005%
Loss on Drying (150 °C, 4 h) ≤0.5%
Residual solvents (Ph. Eur./USP/ICH) Excluded by Manufacturing Process

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge