1.07730.1000
1.07730.5000
1.07730.9025
1.07731.1000
1.07731.5000
1.07731.9025
1.07736.1000
1.07736.2500
1.07736.9025
1.07739.1000
1.07747.1000
1.07747.2500
1.07748.1000
1.07749.1000
1.07749.2500
1.07749.9025
1.07748.2500
1.07730.1000EA
157
EUR
InStock
1.07730.1000
1.07730.5000
1.07730.9025
1.07731.1000
1.07731.5000
1.07731.9025
1.07736.1000
1.07736.2500
1.07736.9025
1.07739.1000
1.07747.1000
1.07747.2500
1.07748.1000
1.07749.1000
1.07749.2500
1.07749.9025
1.07748.2500
Silica gels for TLC and PLC plates, standardised
Chromatographie-Sorbentien
TLC-Sorbentien
Selection of standardised and modified silica gels that are available in many granulations and supplied from one single source, dedicated to chromatography. Standardised silica gels are widely used in separation processes to purify valuable compounds in ton quantities.
- Reliable results and easy sample application
- Variety of packing sizes from 1 to 25 kg
- Economic alternatives to conventional types
* F254: Green fluorescent indicator, H: Without foreign binder, G: With gypsum, P: For preparative work.
Verpackung: Supplied in plastic or tin packaging.