Silica gels for TLC and PLC plates, standardised
Lieferant: MERCK MILLIPORE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.07730.1000 1.07730.5000 1.07730.9025 1.07731.1000 1.07731.5000 1.07731.9025 1.07736.1000 1.07736.2500 1.07736.9025 1.07739.1000 1.07747.1000 1.07747.2500 1.07748.1000 1.07749.1000 1.07749.2500 1.07749.9025 1.07748.2500
1.07730.1000EA 157 EUR InStock
1.07730.1000 1.07730.5000 1.07730.9025 1.07731.1000 1.07731.5000 1.07731.9025 1.07736.1000 1.07736.2500 1.07736.9025 1.07739.1000 1.07747.1000 1.07747.2500 1.07748.1000 1.07749.1000 1.07749.2500 1.07749.9025 1.07748.2500
Silica gels for TLC and PLC plates, standardised
Chromatographie-Sorbentien TLC-Sorbentien
Selection of standardised and modified silica gels that are available in many granulations and supplied from one single source, dedicated to chromatography. Standardised silica gels are widely used in separation processes to purify valuable compounds in ton quantities.
  • Reliable results and easy sample application
  • Variety of packing sizes from 1 to 25 kg
  • Economic alternatives to conventional types
* F254: Green fluorescent indicator, H: Without foreign binder, G: With gypsum, P: For preparative work.
Verpackung: Supplied in plastic or tin packaging.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge