Schnelltests, Formaldehyd-Test, MQuant®, Supelco®
Lieferant: MERCK MILLIPORE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.08028.0001
1.08028.0001EA 205 EUR InStock
1.08028.0001
Schnelltests, Formaldehyd-Test, MQuant®, Supelco®
Umwelt-Prüfkits
Quick tests, visual colour comparison.
  • Results in approximately 5 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
Lieferumfang: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge