Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

(±)-2-Methyltetrahydrofuran, EMPLURA®

Lieferant: MERCK MILLIPORE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

MQ200

1.08292.2500 1.08292.1000 1.08292.4000
1.08292.2500EA 96.8 EUR InStock
1.08292.2500 1.08292.1000 1.08292.4000
(±)-2-Methyltetrahydrofuran, EMPLURA®
(±)-2-Methyltetrahydrofuran
2-Methyltetrahydrofuran (Methyl THF) is a safer and more sustainable alternative to dichloromethane and tetrahydrofuran. It is derived from renewable resources, such as corncobs and sugarcane bagasse.

  • Less solvent consumption due to more efficient extraction and higher reaction yields
  • Lower volatility and higher flash point increase user safety
  • Limited miscibility in water reduces waste stream
  • Reliable availability (independent of petrochemical production)


Formel: C₅H₁₀O
MW: 86.13 g/mol
Siedepunkt: 79
Melting Pt: -136
Flash-Pt: -12
Storage Temperature: Raumtemperatur
MDL Number: MFCD00005367
CAS-Nummer: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3 II
Order Now
Spezifikation Testergebnisse
Hill Formula C₅H₁₀O
Molar Mass 86.13 g/mol
Boiling point 78 - 80 °C
Density 0.85 g/cm³ (20 °C)
Explosion limit 1.2 - 5.7 %(V)
Flash point -12 °C
Ignition temperature 270 °C
Melting Point -136 °C
Vapor pressure 130 hPa (25 °C)
Solubility 130 g/l (25 °C)
LD 50 oral LD50 Rat300 - 2000 mg/kg
LD 50 dermal LD50 Rat2000 mg/kg
Storage class 3 Flammable liquids
WGK WGK 2 water endangering
R Phrase R 11-19-22-38-41
S Phrase S 16-29-33-26-39

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge