Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Titriplex® III -Lösung c (Na₂-EDTA , 2 H₂O) = 0,1 mol/l, Titripur® Reag. Ph. Eur. für die Metalltitration, Supelco®

Lieferant: MERCK MILLIPORE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M-Clarity™ Qualitätsstufe = MQ100

1.08431.1000 1.08431.9010 1.08431.4000 1.08431.1003
1.08431.1000EA 28.88 EUR InStock
1.08431.1000 1.08431.9010 1.08431.4000 1.08431.1003
Titriplex® III -Lösung c (Na₂-EDTA , 2 H₂O) = 0,1 mol/l, Titripur® Reag. Ph. Eur. für die Metalltitration, Supelco®
EDTA Dinatriumsalz
The concentration is determined by volumetric titration and refers to 20°C.
The certified value is traceable to a primary standard from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 682 zinc) by means of volumetric standard zinc (Art. 1.02409). This volumetric standard is certified by the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%. All measurements are carried out in the laboratory of Merck KGaA, Darmstadt, Germany.

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.


Formel: C₁₀H₁₄N₂Na₂O₈
Dichte: 1,02 g/cm3 (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00070672
CAS-Nummer: 139-33-3
EINECS: 205-358-3
Order Now
Spezifikation Testergebnisse
Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge