Präparative Sorbentien, LiChroprep®
Lieferant: MERCK PRODUCTION CHEMICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.09303.0100 1.09303.0500 1.09336.1000 1.09336.9025 1.09362.0250 1.09362.1000 1.09390.1000 1.13900.0250 1.13900.1000 1.13901.0500 1.13905.0250 1.13905.1000 1.09303.9025 1.09303.5000 1.13900.9025 1.13901.9010
1.09303.0100EA 481 EUR InStock
1.09303.0100 1.09303.0500 1.09336.1000 1.09336.9025 1.09362.0250 1.09362.1000 1.09390.1000 1.13900.0250 1.13900.1000 1.13901.0500 1.13905.0250 1.13905.1000 1.09303.9025 1.09303.5000 1.13900.9025 1.13901.9010
Präparative Sorbentien, LiChroprep®
Chromatographie-Sorbentien HPLC-Sorbentien
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge