Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hämatoxylin Monohydrat, Certistain® für die Mikroskopie, Sigma-Aldrich®

Lieferant: MERCK MILLIPORE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


1.15938.0025 1.15938.0100 1.15938.1000
1.15938.0025EA 156 EUR InStock
1.15938.0025 1.15938.0100 1.15938.1000
Hämatoxylin Monohydrat, Certistain® für die Mikroskopie, Sigma-Aldrich®
Hämatoxylin Monohydrat
Hematoxylin-Monohydrat (C.I. 75290) Certistain® is a dye for the specific staining of DNA in the nuclei. It is used in cytology and histology. Nuclei are stained blue, dark violet to black. Hematoxylins are mixed with trivalent positively charged metal salts and build up the so-called hematoxylin lakes, used for the selective staining of nuclei (DNA). Certistain® is the quality group for seleceted high quality dyes that fulfill all quality requirements for IVD and CE registration.


Formel: C₁₆H₁₄O₆·H₂O
MW: 320.3 g/mol
CAS-Nummer: 517-28-2
EINECS: 208-237-3
IVDLogo
Order Now
Spezifikation Testergebnisse
Hill Formula C₁₆H₁₄O₆ * H₂O
Molar Mass 320.29 g/mol
Melting Point 140 °C Elimination of water of crystallisation
Bulk density 500 kg/m³
Solubility (20 °C) slightly soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 2 water endangering

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge