Reagents for non Merck photometers, Spectroquant®, Supelco®
Lieferant: MERCK MILLIPORE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.19254.0001
1.19254.0001EA 46.8 EUR InStock
1.19254.0001
Reagents for non Merck photometers, Spectroquant®, Supelco®
Umwelt-Prüfkits
Spectroquant® test kits are suitable for use only in conjunction with Hach instruments. They do not require any further device-specific calibration.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge