Sulphate CRM, standard solutions, Supelco®
Lieferant: MERCK MILLIPORE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.25050.0100 1.25051.0100 1.25052.0100 1.25053.0100
1.25050.0100EA 49.6 EUR InStock
1.25050.0100 1.25051.0100 1.25052.0100 1.25053.0100
Sulphate CRM, standard solutions, Supelco®
Standards Anionische Standards Sulfat-Standards
Na₂SO₄ in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
The concentrations available are designed for use with Spectroquant® test kits, but are equally applicable to standard methods or for use with other manufacturers' relevant test kits.
Zertifizierungen: Traceable to SRM from NIST and USP.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge