Bromate Standard Solution, CRM traceable to SRM from NIST, 0.1000 mg/l BrO3- in H2O 1 * 100 mL
Lieferant: MERCK MILLIPORE
Bromate Standard Solution, CRM traceable to SRM from NIST, 0.1000 mg/l BrO3- in H2O 1 * 100 mL
PRODUKT
Erweitert / Kompakt
Bromate Standard Solution, CRM traceable to SRM from NIST, 0.1000 mg/l BrO3- in H2O 1 * 100 mL 1.33007.0100 1 * 100 mL
Hersteller-Artikelnummer:: 1.33007.0100
Lokale Artikelnummer:: MERC1.33007.0100
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder