HPLC columns, LiChroCART®, Purospher® STAR RP-8e
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.51432.7220 1.50229.7194 1.50170.7220 1.50228.7194 1.50201.7194 1.50195.7220 1.50173.7220 1.50177.7194 1.50178.7194 1.51444.7194 1.50170.7194 1.50171.7194 1.51431.7220 1.51432.7194 1.50201.7220 1.50233.7220 1.50233.7194 1.50234.7194 1.50236.7194 1.50236.7220 1.50302.7194 1.50302.7220 1.51431.7194 1.51442.7194 1.51442.7220 1.51443.7194 1.51445.7194 1.50174.7194 1.50179.7194 1.51448.7194 1.51448.7220 1.50174.7220 1.50177.7220
1.51432.7220EA 0 EUR InStock
1.51432.7220 1.50229.7194 1.50170.7220 1.50228.7194 1.50201.7194 1.50195.7220 1.50173.7220 1.50177.7194 1.50178.7194 1.51444.7194 1.50170.7194 1.50171.7194 1.51431.7220 1.51432.7194 1.50201.7220 1.50233.7220 1.50233.7194 1.50234.7194 1.50236.7194 1.50236.7220 1.50302.7194 1.50302.7220 1.51431.7194 1.51442.7194 1.51442.7220 1.51443.7194 1.51445.7194 1.50174.7194 1.50179.7194 1.51448.7194 1.51448.7220 1.50174.7220 1.50177.7220
HPLC columns, LiChroCART®, Purospher® STAR RP-8e
Chromatographiesäulen
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge