LiChroCART®, Superspher® 100 Si HPLC Columns
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.50170.7143 1.50195.7143 1.50201.7143 1.50228.7143 1.50229.7143 1.50233.7143 1.50234.7143 1.50236.7143 1.50302.7143 1.51431.7143 1.50174.7143 1.51432.7143 1.51442.7143 1.51443.7143 1.51444.7143 1.51445.7143 1.50171.7143 1.51448.7143 1.50173.7143 1.50175.7143 1.50177.7143 1.50178.7143 1.50179.7143
1.50170.7143EA 0 EUR InStock
1.50170.7143 1.50195.7143 1.50201.7143 1.50228.7143 1.50229.7143 1.50233.7143 1.50234.7143 1.50236.7143 1.50302.7143 1.51431.7143 1.50174.7143 1.51432.7143 1.51442.7143 1.51443.7143 1.51444.7143 1.51445.7143 1.50171.7143 1.51448.7143 1.50173.7143 1.50175.7143 1.50177.7143 1.50178.7143 1.50179.7143
LiChroCART®, Superspher® 100 Si HPLC Columns
Chromatographiesäulen
Stainless steel
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge