Drucken

Kaliumchlorid 1 mol/l (1 N) (20°C), Titripur®, Supelco®

Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

1.60331.1003
1.60331.1003EA 97.2 EUR
1.60331.1003
Kaliumchlorid 1 mol/l (1 N) (20°C), Titripur®, Supelco®
Potassium chloride

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: KCl
MW: 74.55 g/mol
Siedepunkt: 1420
Melting Pt: 773
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011360
CAS-Nummer: 7447-40-7
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR