BCIP/NBT Kit
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10003
10003.EA 238 EUR InStock
10003.
BCIP/NBT Kit
BCIP/NBT
This kit contains 100 mg each of BCIP sodium salt (10001) and the oxidant NBT (Nitro blue tetrazolium, 10008).
BCIP is the most widely used chromogenic phosphatase substrate, which forms a dark blue precipitate on enzymatic hydrolysis. It is often used with the oxidant NBT (nitro blue tetrazolium chloride), which facilitates the precipitation. BCIP/NBT produces a dark blue/violet precipitate product that can be mounted with permanent or aqueous mounting medium.
Stored desiccated at ≤ 4 °C upon receipt. Protect components from light, especially when in solution.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge