10020.ND
10020.NF
10020.NDEA
160
EUR
InStock
10020.ND
10020.NF
Cäsiumhydroxid 50% (w/w) in wässriger Lösung , 99.9% (Metall-Basis)
Cesium hydroxide
Purity: 99.9%
CAS No.: 21351-79-1
Molecular Formula: CsOH
Formula Weight: 149.91
Physical Form: Liquid
Density: 1.725
Appearance: Colourless
Odour: Odourless
UN No.: 2681
MDL No.: MFCD00010964