1-(4-Nitrophenylazo)-2-naphthol, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M-Clarity™ Qualitätsstufe = MQ100
100994-25G
100994-25GEA 129 EUR InStock
100994-25G
1-(4-Nitrophenylazo)-2-naphthol, Sigma-Aldrich®
1-(4-Nitrophenylazo)-2-naphthol
Para red is a biological stain. It is used as a fat soluble chemical dye and food additive. It is a banned colorant and a number of analytical techniques have been developed to detect it in food. Para Red may be used to make standards for analysis.
Formel: C₁₆H₁₁N₃O₃
Melting Pt: 248…252 °C (Lit.)
MDL Number: MFCD00003908
CAS-Nummer: 6410-10-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9da56c7639057c375e66e20eff541294d8908ec4&vid=9da56c7639057c375e66e20eff541294d8908ec4&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge