FectoVIR®-LV Transfection Reagents
Lieferant: POLYPLUS TRANSFECTION
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
101000187 101000188 101000189
101000187.EA 294 EUR InStock
101000187. 101000188. 101000189.
FectoVIR®-LV Transfection Reagents
Transfektionsreagenzien
Transfection reagents for lentiviral vector production from small to large scale.
  • Productivity: Reach highest lentiviral vectors titers in suspension systems
  • Cost-effectiveness: Reduce your cost per batch with high titers and low DNA consumption
  • Scalability: Produce at large scale with low complexation volume and long complex stability
  • Time saving: Use polyplus DOE service to optimize your process in a record time
FectoVIR®-LV is the next generation of transfection reagent, free of animal component, designed to improve Lentiviral vector productivity in HEK-293 cell systems.
FectoVIR®-LV is made for large scale manufacturing with a reduction of the complexation volume and increased complex stability.
The FectoVIR-LV benefits allow to increase the number of doses produced per batch, while decreasing LVV manufacturing costs.
Zertifizierungen: ISO 9001 facility certifications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge