101000187
101000188
101000189
101000187.EA
294
EUR
InStock
101000187.
101000188.
101000189.
FectoVIR®-LV Transfection Reagents
Transfektionsreagenzien
Transfection reagents for lentiviral vector production from small to large scale.
- Productivity: Reach highest lentiviral vectors titers in suspension systems
- Cost-effectiveness: Reduce your cost per batch with high titers and low DNA consumption
- Scalability: Produce at large scale with low complexation volume and long complex stability
- Time saving: Use polyplus DOE service to optimize your process in a record time
FectoVIR®-LV is the next generation of transfection reagent, free of animal component, designed to improve Lentiviral vector productivity in HEK-293 cell systems.
FectoVIR®-LV is made for large scale manufacturing with a reduction of the complexation volume and increased complex stability.
The FectoVIR-LV benefits allow to increase the number of doses produced per batch, while decreasing LVV manufacturing costs.
Zertifizierungen: ISO 9001 facility certifications.