unter Argon abgefüllt
10103.22
10103.36
10103.A1
10103.22EA
230
EUR
InStock
10103.22
10103.36
10103.A1
Barium 99+% (Metall-Basis), Stäbe (max. 0,8% Si), Ø 22 mm (0.9 in)
Barium
Purity: 99+%
CAS No.: 7440-39-3
Physical Form: ≈7.3cm/100g, Packaged under argon
Appearance: Grey
Odour: Odourless
UN No.: 1400
MDL No.: MFCD00134031
Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps