10147.A3
10147.09
10147.22
10147.36
10147.A3EA
1155
EUR
InStock
10147.A3
10147.09
10147.22
10147.36
Chrom 99 Metals basis, powder -100 mesh
Chromium
Purity: 99%
CAS No.: 7440-47-3
Physical Form: Apparent density 2.0-3.0g/cm3
Appearance: Silver
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00010944