10535.03
10535.03EA
198
EUR
InStock
10535.03
Potassium tetracyanopalladate(II) hydrate
Potassium tetracyanopalladate(II) hydrate
CAS No.: 14516-46-2
Molecular Formula: K2[Pd(CN)4].xH2O
Formula Weight: 288.68(anhy)
Physical Form: Crystalline
Appearance: White
MDL No.: MFCD00150431