10598.18
10598.09
10598.90
10598.18EA
382
EUR
InStock
10598.18
10598.09
10598.90
Antimon ≥99,9999% (Metall-Basis), polykristalliner Brocken, Puratronic® 2-8 mm (0.08-0.3 in)
Antimony
Purity: 99.9999%
CAS No.: 7440-36-0
Odour: Odourless
MDL No.: MFCD00134030