10646.14
10646.22
10646.36
10646.14EA
74.2
EUR
InStock
10646.14
10646.22
10646.36
Bariumchlorid Dihydrat ≥99,997% (Metall-Basis), Puratronic®
Bariumchlorid Dihydrat
Purity: 99.997%
CAS No.: 10326-27-9
Molecular Formula: BaCl2.2H2O
Formula Weight: 244.28 (208.25anhy)
Physical Form: Powder
Density: 3.097
Appearance: White
Odour: Odourless
Melting Point: 113° -2H2O
Merck Reference: 14,971
UN No.: 1564
MDL No.: MFCD00149154