10647.14
10647.22
10647.36
10647.88
10647.14EA
132
EUR
InStock
10647.14
10647.22
10647.36
10647.88
Bariumfluorid ≥99,99% (Metall-Basis), Puratronic®
Barium fluoride
Purity: 99.99%
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: Powder
Density: 4.83
Refractive Index: 1.4741
Odour: Odourless
Melting Point: 1353°
Boiling Point: 2260°
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450