10648.06
10648.14
10648.22
10648.36
10648.88
10648.06EA
33.2
EUR
InStock
10648.06
10648.14
10648.22
10648.36
10648.88
Bariumnitrat ≥99,999% (Metall-Basis), Puratronic®
Barium nitrate
Purity: 99.999%
CAS No.: 10022-31-8
Molecular Formula: BaN2O6
Formula Weight: 261.35
Physical Form: Crystalline
Density: 3.24
Appearance: White
Melting Point: 592° dec.
Merck Reference: 14,983
UN No.: 1446
MDL No.: MFCD00003442