Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bismut(III)nitrat, Hyrdrat ≥99,999% (Metall-Basis), Puratronic®

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

10657.14 10657.22
10657.14EA 99.2 EUR InStock
10657.14 10657.22
Bismut(III)nitrat, Hyrdrat ≥99,999% (Metall-Basis), Puratronic®
Bismut(III)nitrat
It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.

Purity: 99.999% CAS No.: 10361-44-1 Molecular Formula: Bi(NO3)3.xH2O Formula Weight: 394.99(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Colourless to white Melting Point: 30° dec. Merck Reference: 14,1271 UN No.: 1477 MDL No.: MFCD00003460


Formel: Bi(NO₃)₃·xH₂O
Siedepunkt: 75…80 °C (1013 hPa)
Melting Pt: 30 °C (Zersetzung)
Dichte: 2,736 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003460
CAS-Nummer: 10361-44-1
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Merck Index: 14,1271
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge