10729.09
10729.18
10729.09EA
178
EUR
InStock
10729.09
10729.18
Blei(IV)oxid ≥99,995% (Metall-Basis), Puratronic®
Blei(IV)oxid
Purity: 99.995%
CAS No.: 1309-60-0
Molecular Formula: PbO2
Formula Weight: 239.19
Physical Form: Powder
Density: 9.38
Melting Point: 290° dec.
Merck Reference: 14,5407
UN No.: 1872
MDL No.: MFCD00011165