11094.22
11094.36
11094.09
11094.22EA
57.1
EUR
InStock
11094.22
11094.36
11094.09
Vanadium(V)oxid ≥99,6%
Vanadium(V)oxid
Purity: 99.6% min
CAS No.: 1314-62-1
Molecular Formula: V2O5
Formula Weight: 181.88
Physical Form: -10 Mesh
Density: 3.357
Melting Point: 690°
Boiling Point: 1750° dec.
Merck Reference: 14,9921
UN No.: 2862
MDL No.: MFCD00011457