Safety goggles, Vanguard UDC
Lieferant: UNIVET
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6X3.00.00.00
111-2516EA 168 EUR InStock
111-2516
Safety goggles, Vanguard UDC
Vollsichtbrillen
6X3 goggles are designed with the intention of combining a higher level of protection and innovative style. Equipped with a panoramic lens, they offer excellent peripheral vision and can be used comfortably with any corrective eyewear.
  • Panoramic lens can be worn over prescription glasses
  • Exclusive Vanguard UDC coating
  • Possibility to quickly replace the lens with a special release
  • Facial support compatible with half masks
  • Indirect ventilation system ensures certified protection against drops and splashes
  • Impact resistance at 432 km/h, even at extreme temperatures
The design of the face support ensures proper seating for respiratory masks and half masks, and in the use of the facial protection, providing additional protection to the operator’s face.
Weight: 78 g
Zertifizierungen: Category III° PPE
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge