Coverlite™ Safety spectacle, Stealth™
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ASA940-061-300 ASA940-063-000 ASA940-0A3-000
111-2597EA 9.8 EUR InStock
111-2597 111-2598 111-2599
Coverlite™ Safety spectacle, Stealth™
Schutzbrillen
Stylish lightweight overspec for use with prescription eyewear.
  • Stylish overspec
  • Optically perfect
  • Pantoscopic soft grip arms
  • Flexible frame
  • Ultra lightweight
The stylish and spacious design of the Stealth™ Coverlite™ allows them to comfortably fit over prescription eyewear while maintaining a modern look, unlike traditional bulkier overspectacles. The frame is flexible and springy so that they can fit any head size and create a secure fit. The robust, low base, minimum curve, optical Class 1 lens provides optimal vision to the wearer.
One of the lightest overspectacles on the market, weighing just 34 g. Two levels of lens coating are available; K class anti-scratch or K&N class anti-scratch and anti-mist combined.
Zertifizierungen: Conforms to EN 166
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge