Full face masks with Typhoon™ valve, Force®10
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BPB003-004-000 BPB003-104-000 BPB003-204-000
111-2600EA 267 EUR InStock
111-2600 111-2601 111-2602
Full face masks with Typhoon™ valve, Force®10
Atemschutzmasken Mehrweg-Atemschutzmasken Vollmasken
The Force®10 Typhoon™ is an extremely lightweight, easy to maintain, full face mask with panoramic visor. With 3 available sizes (S,M,L) and the ability to fit PressToCheck™ filters, the user can feel reassured of an excellent fit.
  • Available in Small, Medium, or Large sizes for a perfect fit
  • Compatible with PressToCheck™ Filters (not included)
  • Typhoon™ exhalation valve for reduced breathing resistance
  • Polycarbonate lens, ABS buckles
  • Engineered for easy maintenance
  • One of the lightest full face masks on the market
The Force®10 Typhoon™ has been meticulously engineered to incorporate a small number of key components. Disassembly, cleaning, and maintenance are effortless with replacement parts available if necessary. Compatible with both JSP Classic filters and the PressToCheck™ filters, which allow for daily face fit reassurance.
Comfort is a key feature of this ultra-lightweight mask, as the web-like structure of the harness cradles the users head, ensuring wearer comfort throughout the working day. The addition of different sizing options allows for a more tailored fit and greater comfort.
The unique Typhoon™ valve has been designed to reduce breathing resistance, heat, and moisture build-up. The valve is unaffected by any moisture, and therefore can be used with the same performance under very low temperatures (−40 °C tested).
Zertifizierungen: Facepiece EN136. Gas Filter Performance EN14387. Particulate Filter Performance EN143. Flammability EN136.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge