11136.18
11136.30
11136.18EA
230
EUR
InStock
11136.18
11136.30
Zink(II)nitrat Hexahydrat ≥99,998% (Metall-Basis), Puratronic®
Zink(II)nitrat Hexahydrat
Purity: 99.998%
CAS No.: 10196-18-6
Molecular Formula: N2O6Zn.6H2O
Formula Weight: 297.47 (189.38anhy)
Physical Form: Crystalline Aggregates
Density: 2.07
Appearance: White
Odour: Odourless
Melting Point: ca 36°
Merck Reference: 14,10144
UN No.: 1514
MDL No.: MFCD00149889