Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Kaliumiodid 99,0-101,5%, GenAR® USP, Macron Fine Chemicals™

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

C1115.0500EA 0 EUR InStock
C1115.0500 1115-04 1115-05 1115-20 1115-24 1115-65
Kaliumiodid 99,0-101,5%, GenAR® USP, Macron Fine Chemicals™
Kaliumiodid


Formel: Kl
MW: 166 g/mol
Siedepunkt: 1330
Melting Pt: 681 °C
MDL Number: MFCD00011405
CAS-Nummer: 7681-11-0
Merck Index: 13,07727
Order Now
Spezifikation Testergebnisse
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical 99.0 - 101.5 %
USP - Assay (KI)(U.S.P.) 99.0 - 101.5 %
USP - Identification (Potassium) Passes Test
USP - Identification (Iodide) Passes Test
USP - Loss on Drying at 105°C ≤0.50 %
USP - Iodate (IO₃) ≤4 mg/kg
USP - Thiosulfate and Barium Passes Test
USP - Alkalinity Passes Test
USP - Nitrate, Nitrite, and Ammonia Passes Test
EP/BP - Assay (Dried Basis) 99.0 - 100.5 %
EP/BP - Identificaton A (Iodides) Passes Test
EP/BP - Identification B (Potassium) Passes Test
EP/BP - Alkalinity Passes Test
EP/BP - Appearance (white, granular) Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Iodates Passes Test
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Thiosulphates Passes Test
EP/BP - Iron (Fe) ≤20 ppm
EP/BP - Loss on Drying ≤1.0 %
Free Alkali, For Information Only = 10 ppm
FCC - Assay (KI)(F.C.C.) 99.0 - 101.5 %
FCC - Identification (Potassium) Passes Test
FCC - Identification (Iodide) Passes Test
FCC - Identification B (FCC) Passes Test
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying at 105°C ≤0.50 %
FCC - Iodate (IO₃) ≤4 mg/kg
FCC - Thiosulfate and Barium Passes Test
FCC - Nitrate, Nitrite, and Ammonia Passes Test
No Class 1,2,3 or other solvents are used or produced in the Passes Test
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge