Chemical and cut resistant gloves, Showa 3416
Lieferant: Showa
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3416-08 3416-10 3416-11 3416-09
112-5784EA 273 EUR InStock
112-5784 112-5786 112-5787 112-5785
Chemical and cut resistant gloves, Showa 3416
Handschuhe
Constructed with a seamless knit liner for maximum comfort and endurance, this fully coated neoprene gauntlet provides ANSI level A4 and EN level 5 cut protection and protection against acids, caustics, solvents, grease and oil.
  • Seamless liner made from cut resistant engineered fibre
  • Fully coated
  • Flexible coating with maximum comfort and protection
  • Rough particle finish
  • Gauntlet cuff for protection of the forearm
Recommended industries: Transport, steel erecting, mechanical, metallurgy, petrochemical, automotive.
Zertifizierungen: EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5; Cat. III
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge