Safety helmet, uvex pheos planet B-S-WR
Lieferant: UVEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9772.043
112-6185EA 28 EUR InStock
112-6185
Safety helmet, uvex pheos planet B-S-WR
Schutzhelme Schutzhelme
Lightweight helmets made from HDPE.
  • Suspension harness with wheel ratchet system for variable width adjustment
  • Safety helmet with helmet torch slot on the front
  • Euroslot adapters on the side (30 mm) for attaching earmuffs and the uvex pheos visor system
  • The shortened brim ensures unrestricted upwards vision
  • Six-point textile band attachment ensures excellent fit and comfort
  • Three adjustable air vents for maximum ventilation
Ideal for industry, road construction works, sewage and pipeline construction, concrete/construction work, brickwork, stonemasonry, plasterwork, demolition/dismantling work, bridge/tunnel construction, high street construction and metalwork.
Adjustable size range: 520 to 610 mm
Zertifizierungen: Meets EN 397 standard and additional requirements for very low temperatures (−30 °C) and molten metal (MM) splash.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge