11237.14
11237.22
11237.14EA
80.5
EUR
InStock
11237.14
11237.22
Praseodymtrichlorid Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Praseodymtrichlorid Hydrat
Purity: 99.9%
CAS No.: 19423-77-9
Molecular Formula: PrCl3.xH2O
Formula Weight: 247.27(anhy)
Physical Form: Crystalline Aggregates
Odour: Odourless
Merck Reference: 14,7712
MDL No.: MFCD00149828