11268.18
11268.30
11268.A1
11268.18EA
91.3
EUR
InStock
11268.18
11268.30
11268.A1
Lanthan(III)chlorid Heptahydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lanthan(III)chlorid Heptahydrat
Purity: 99.99%
CAS No.: 10025-84-0
Molecular Formula: LaCl3.7H2O
Formula Weight: 371.37 (245.27anhy)
Physical Form: Wet Crystalline Aggregates
Odour: Odourless
Merck Reference: 14,5363
MDL No.: MFCD00149756