11287.06
11287.14
11287.22
11287.06EA
93.6
EUR
InStock
11287.06
11287.14
11287.22
Gadolinium(III)chlorid Hexahydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Gadolinium(III)chlorid Hexahydrat
Purity: 99.99%
CAS No.: 13450-84-5
Molecular Formula: GdCl3.6H2O
Formula Weight: 371.70 (263.61anhy)
Physical Form: Crystalline Aggregates
Density: 2.424
Appearance: White
Odour: Characteristic
Merck Reference: 14,4326
MDL No.: MFCD00149732