Industrieller Klettersitz
Lieferant: HONEYWELL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1007056
113-1250EA 442 EUR InStock
113-1250
Industrieller Klettersitz
Fallschutz Auffanggurte
This detachable climbers seat offers a comfortable supported position for the operator during prolonged periods suspended at height. It is made from marine ply covered with closed cell foam and abrasion resistant PVC coated nylon.
  • Padded seat with waterproof PVC coated nylon covering
  • Adjustable straps for perfect fit
  • Supplied with two connectors and convenient ring attachments for accessories
Please note: Must only be used with an appropriate safety harness.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge